siRNA Details
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106711.1 | NP_001100181.1 | 2, 3 | 284 | ||
XM_006239798.2 | XP_006239860.1 | 2, 3 | 407 | |||
GenBank mRNA | BC166997.1 | - | 2, 3 | 325 | 8365241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
