siRNA Details
Gene Symbol: Nt5c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011982.1 | NP_001011982.1 | 3 | 250 | ||
XM_006239888.2 | XP_006239950.1 | 4 | 452 | |||
XM_006239889.2 | XP_006239951.1 | 4 | 452 | |||
XM_006239890.2 | XP_006239952.1 | 4 | 329 | |||
XM_006239891.2 | XP_006239953.1 | 3 | 278 | |||
XM_006239892.2 | XP_006239954.1 | 3 | 286 | |||
GenBank mRNA | BC079437.1 | - | 3 | 250 | 7115294 |
