siRNA Details
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106718.2 | NP_001100188.1 | 10, 11 | 1023 | ||
XM_008764584.1 | XP_008762806.1 | 10, 11 | 1572 | |||
XM_008764585.1 | XP_008762807.1 | 9, 10 | 925 | |||
GenBank mRNA | BC158700.1 | - | 10, 11 | 1023 | 9028479 |
