siRNA Details
Gene Symbol: Pqlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034952.1 | NP_001030124.1 | 6 | 667 | ||
XM_008764587.1 | XP_008762809.1 | 5 | 869 | |||
GenBank mRNA | BC081879.1 | - | 4 | 1822 | 7108131 | |
CK477351.1 | - | 6 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
