siRNA Details
Gene Symbol: Iah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100540.1 | NP_001094010.1 | 1, 2 | 92 | ||
GenBank mRNA | BC168771.1 | - | 1, 2 | 92 | 6919650 | |
FQ220941.1 | - | 1, 2 | 88 | |||
FQ225724.1 | - | 1, 2 | 117 | |||
FQ231316.1 | - | 1, 2 | 117 | |||
FQ232084.1 | - | 1, 2 | 122 | |||
FQ235324.1 | - | 1, 2 | 117 |

More Information
Other Species Targeted

Gene Ontology Categories:
