siRNA Details
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013097.2 | NP_001013115.1 | 6, 7 | 712 | ||
NM_001286938.1 | NP_001273867.1 | 6, 7 | 712 | |||
GenBank mRNA | BC091209.1 | - | 6, 7 | 687 | 7381386 |
