siRNA Details
Gene Symbol: Lamb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750137.3 | XP_003750185.1 | 33 | 5571 | ||
XM_006239996.2 | XP_006240058.1 | 34 | 5440 | |||
XM_006239997.2 | XP_006240059.1 | 34 | 5449 |
