siRNA Details
Gene Symbol: Nkx1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_234082.7 | XP_234082.6 | 2 | 1165 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nkx1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_234082.7 | XP_234082.6 | 2 | 1165 |