siRNA Details
Gene Symbol: Arhgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047869.1 | NP_001041334.1 | 1 | 3172 | ||
XM_008764656.1 | XP_008762878.1 | 1 | 3192 | |||
GenBank mRNA | AY387084.1 | - | 1 | 3172 |
