siRNA Details
Gene Symbol: Eapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134987.1 | NP_001128459.1 | 6 | 690 | ||
XM_006240100.2 | XP_006240162.1 | 6 | 731 | |||
GenBank mRNA | BC168880.1 | - | 6 | 690 | 9099474 |

More Information
Other Species Targeted

Gene Ontology Categories:
