siRNA Details
Gene Symbol: ZNF142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105537.2 | NP_001099007.1 | 9 | 5021 | ||
NR_073599.1 | 10 | 5330 | ||||
NR_073600.1 | 10 | 5163 | ||||
XM_006712730.3 | XP_006712793.1 | 10 | 5696 | |||
XM_011511784.2 | XP_011510086.1 | 10 | 5791 | |||
XM_011511785.2 | XP_011510087.1 | 10 | 5716 | |||
XM_011511786.2 | XP_011510088.1 | 10 | 5621 | |||
XM_011511787.2 | XP_011510089.1 | 9 | 5616 | |||
XM_011511788.2 | XP_011510090.1 | 9 | 5521 | |||
XM_011511789.2 | XP_011510091.1 | 9 | 5189 | |||
XM_017004871.1 | XP_016860360.1 | 9 | 5772 | |||
XM_017004872.1 | XP_016860361.1 | 8 | 5172 | |||
GenBank mRNA | AK095027.1 | - | 3 | 1674 | ||
BC033631.1 | - | 9 | 5744 | |||
BC053849.2 | - | 10 | 5349 | |||
D87073.1 | - | 9 | 5019 |

More Information
Other Species Targeted

Gene Ontology Categories:
