siRNA Details
Gene Symbol: Aldoart2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013943.1 | NP_001013965.1 | 1 | 783 | ||
GenBank mRNA | BC079243.1 | - | 1 | 783 | 7132739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
