siRNA Details
Gene Symbol: Brms1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106731.1 | NP_001100201.1 | 1131 | |||
GenBank mRNA | BC162034.1 | - | 1038 | 7096422 |
Gene Symbol: LOC100910898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750158.2 | XP_003750206.1 | 10 | 1057 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001037756, AK003055, AK155742, AK154678, BC034885.
Gene Ontology Categories:
