siRNA Details
Gene Symbol: Klhl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106735.1 | NP_001100205.1 | 5 | 1741 | ||
XM_006240135.2 | XP_006240197.1 | 5 | 2672 |

More Information
Other Species Targeted

