siRNA Details
Gene Symbol: Pole2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169108.1 | NP_001162579.1 | 5 | 431 | ||
XM_006240154.2 | XP_006240216.1 | 5 | 446 | |||
XM_008764696.1 | XP_008762918.1 | 8 | 1230 | |||
XM_008764697.1 | XP_008762919.1 | 7 | 1040 | |||
XM_008764698.1 | XP_008762920.1 | 8 | 1169 | |||
XM_008764699.1 | XP_008762921.1 | 8 | 1253 | |||
XM_008764700.1 | XP_008762922.1 | 8 | 1139 | |||
XM_008764701.1 | XP_008762923.1 | 8 | 1033 | |||
XM_008764702.1 | XP_008762924.1 | 8 | 1129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
