siRNA Details
Gene Symbol: Pole2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169108.1 | NP_001162579.1 | 12 | 986 | ||
XM_006240154.2 | XP_006240216.1 | 12 | 1001 | |||
XM_008764696.1 | XP_008762918.1 | 15 | 1785 | |||
XM_008764697.1 | XP_008762919.1 | 14 | 1595 | |||
XM_008764698.1 | XP_008762920.1 | 15 | 1724 | |||
XM_008764699.1 | XP_008762921.1 | 15 | 1808 | |||
XM_008764700.1 | XP_008762922.1 | 15 | 1694 | |||
XM_008764701.1 | XP_008762923.1 | 15 | 1588 | |||
XM_008764702.1 | XP_008762924.1 | 15 | 1684 | |||
GenBank mRNA | FQ231562.1 | - | 6 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
