siRNA Details
Gene Symbol: ZNF143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282656.1 | NP_001269585.1 | 11 | 1126 | ||
NM_001282657.1 | NP_001269586.1 | 10 | 1036 | |||
NM_003442.5 | NP_003433.3 | 11 | 1129 | |||
XM_005253121.1 | XP_005253178.1 | 11 | 1077 | |||
XM_005253122.1 | XP_005253179.1 | 11 | 1102 | |||
XM_005253127.1 | XP_005253184.1 | 12 | 1155 | |||
XM_011520349.2 | XP_011518651.1 | 11 | 1848 | |||
XM_017018252.1 | XP_016873741.1 | 10 | 1013 | |||
XM_017018253.1 | XP_016873742.1 | 10 | 1010 | |||
XM_017018254.1 | XP_016873743.1 | 11 | 1114 | |||
XM_017018255.1 | XP_016873744.1 | 11 | 1204 | |||
XM_017018256.1 | XP_016873745.1 | 10 | 1016 | |||
GenBank mRNA | AF071771.1 | - | 8 | 749 | ||
AK291133.1 | - | 11 | 1082 | |||
AK297214.1 | - | 10 | 981 | |||
AK313330.1 | - | 11 | 1115 | |||
BC020219.1 | - | 11 | 1128 | 4902668 |
