siRNA Details
Gene Symbol: Tex21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025658.1 | NP_001020829.1 | 8 | 1659 | ||
XM_006240226.2 | XP_006240288.1 | 10 | 1862 | |||
XM_006240227.2 | XP_006240289.1 | 10 | 1906 | |||
XM_008764743.1 | XP_008762965.1 | 10 | 1900 | |||
XM_008764744.1 | XP_008762966.1 | 9 | 1777 | |||
XM_008764745.1 | XP_008762967.1 | 8 | 1382 | |||
GenBank mRNA | BC097422.1 | - | 8 | 1659 | 7454996 |
