siRNA Details
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191818.1 | NP_001178747.1 | 8 | 833 | ||
XM_008764746.1 | XP_008762968.1 | 9 | 1053 | |||
XM_008764747.1 | XP_008762969.1 | 10 | 1034 | |||
XM_008764748.1 | XP_008762970.1 | 9 | 1052 | |||
XM_008764749.1 | XP_008762971.1 | 8 | 882 | |||
XM_008764750.1 | XP_008762972.1 | 9 | 1052 | |||
XM_008764751.1 | XP_008762973.1 | 8 | 1299 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001267626, NM_058212, NM_001267625, AK039011, BC048572, and others.
Gene Ontology Categories:
