siRNA Details
Gene Symbol: Arel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106744.1 | NP_001100214.1 | 3, 4 | 405 | ||
XM_006240333.2 | XP_006240395.1 | 4, 5 | 1391 | |||
XM_008764754.1 | XP_008762976.1 | 3, 4 | 666 | |||
XM_008764755.1 | XP_008762977.1 | 5, 6 | 1498 |

More Information
Other Species Targeted

Gene Ontology Categories:
