siRNA Details
Gene Symbol: Zdhhc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039325.1 | NP_001034414.1 | 2 | 510 | ||
GenBank mRNA | AY886537.1 | - | 2 | 510 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001080943, BC128021.
Gene Ontology Categories:
