siRNA Details
Gene Symbol: Ttc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106752.1 | NP_001100222.1 | 6, 7 | 729 | ||
XM_006240422.2 | XP_006240484.1 | 8, 9 | 817 | |||
XM_006240423.2 | XP_006240485.1 | 7, 8 | 786 | |||
XM_006240424.2 | XP_006240486.1 | 8, 9 | 795 | |||
XM_008764762.1 | XP_008762984.1 | 6, 7 | 575 | |||
GenBank mRNA | BC161880.1 | - | 7, 8 | 761 | 7126409 |

More Information
Other Species Targeted

Gene Ontology Categories:
