siRNA Details
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011984.1 | NP_001011984.1 | 6 | 851 | ||
XM_006240464.2 | XP_006240526.1 | 4 | 1496 | |||
GenBank mRNA | BC085882.1 | - | 6 | 851 | 7191983 |
