siRNA Details
Gene Symbol: Ifi27l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206846.2 | NP_996728.1 | 4 | 262 | ||
GenBank mRNA | BC166870.1 | - | 4 | 262 | 6922110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
