siRNA Details
Gene Symbol: Serpina6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009663.1 | NP_001009663.1 | 4 | 1064 | ||
GenBank mRNA | BC088300.1 | - | 4 | 1064 | 7323342 | |
FQ209415.1 | - | 4 | 1109 | |||
FQ211289.1 | - | 4 | 1079 | |||
FQ218294.1 | - | 4 | 1087 | |||
FQ218398.1 | - | 4 | 1074 | |||
FQ219289.1 | - | 4 | 1139 |
