siRNA Details
Gene Symbol: Slc25a47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001509.1 | NP_001001509.1 | 4 | 285 | ||
GenBank mRNA | AY603424.1 | - | 4 | 285 | ||
BC089874.1 | - | 4 | 289 | 7325453 | ||
FQ210765.1 | - | 4 | 345 |
