siRNA Details
Gene Symbol: Cinp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106758.2 | NP_001100228.1 | 4 | 394 | ||
GenBank mRNA | BC158883.1 | - | 4 | 394 | 7937483 |

More Information
Other Species Targeted

Gene Ontology Categories:
