siRNA Details
Gene Symbol: Exoc3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106759.2 | NP_001100229.1 | 2 | 343 | ||
XM_006240572.2 | XP_006240634.1 | 2 | 343 | |||
XM_006240573.2 | XP_006240635.1 | 2 | 301 | |||
XM_006240574.2 | XP_006240636.1 | 2 | 2721 | |||
XM_006240575.2 | XP_006240637.1 | 2 | 447 | |||
XM_006240576.2 | XP_006240638.1 | 2 | 110 | |||
GenBank mRNA | BC166426.1 | - | 2 | 343 | 7100899 | |
FQ230530.1 | - | 2 | 362 |
