siRNA Details
Gene Symbol: Tdrd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764983.1 | XP_008763205.1 | 15 | 1711 | ||
XM_008764984.1 | XP_008763206.1 | 15 | 1711 | |||
XM_008764985.1 | XP_008763207.1 | 15 | 1711 | |||
XM_008764986.1 | XP_008763208.1 | 15 | 1711 | |||
XM_008764987.1 | XP_008763209.1 | 15 | 1711 | |||
XM_008764988.1 | XP_008763210.1 | 15 | 1482 | |||
GenBank mRNA | BC087164.1 | - | 8 | 603 |
