siRNA Details
Gene Symbol: Ighg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088271.1 | - | 863 | 7379698 | ||
BC092584.1 | - | 881 | 7384581 | |||
BC095846.1 | - | 867 | 7388075 |
Gene Symbol: IgG-2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088240.1 | - | 6 | 805 | 7365825 | |
BC088423.1 | - | 6 | 877 | 7123713 | ||
BC091257.1 | - | 806 | 7375893 | |||
BC091272.1 | - | 808 | 7382505 | |||
L22652.1 | - | 7 | 874 | |||
L22654.1 | - | 888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
