siRNA Details
Gene Symbol: Sp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191774.1 | NP_001178703.1 | 2 | 531 | ||
XM_008765001.1 | XP_008763223.1 | 2 | 617 | |||
XM_008765002.1 | XP_008763224.1 | 2 | 540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
