siRNA Details
Gene Symbol: Ctps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134873.1 | NP_001128345.1 | 13 | 1403 | ||
XM_006238781.2 | XP_006238843.1 | 13 | 1396 | |||
GenBank mRNA | BC161954.1 | - | 13 | 1381 | 9027103 | |
FQ210258.1 | - | 12 | 1311 | |||
FQ217494.1 | - | 13 | 1361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
