siRNA Details
Gene Symbol: Rdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199208.1 | NP_954678.1 | 883 | |||
GenBank mRNA | BC062000.1 | - | 850 | 5599919 | ||
BC079153.1 | - | 3 | 707 | 7128686 | ||
U33500.1 | - | 883 |
Gene Symbol: LOC100365958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241512.2 | XP_006241574.1 | 3 | 861 |
