siRNA Details
Gene Symbol: Camkmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134463.1 | NP_001127935.1 | 2, 3 | 366 | ||
GenBank mRNA | BC159411.1 | - | 2, 3 | 366 | 8367183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
