siRNA Details
Gene Symbol: Cdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013103.2 | NP_001013121.2 | 4 | 457 | ||
GenBank mRNA | CB611861.1 | - | 4 | 460 |

More Information
Other Species Targeted

Gene Ontology Categories:
