siRNA Details
Gene Symbol: Abca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207598.1 | NP_997481.1 | 42 | 5858 | ||
XM_006240871.2 | XP_006240933.1 | 43 | 6212 | |||
XM_006240872.2 | XP_006240934.1 | 43 | 6109 | |||
XM_006240873.2 | XP_006240935.1 | 43 | 6100 | |||
XM_006240874.2 | XP_006240936.1 | 43 | 5997 | |||
XM_006240875.2 | XP_006240937.1 | 41 | 5951 | |||
GenBank mRNA | AB097814.1 | - | 42 | 5858 |

More Information
Other Species Targeted

Gene Ontology Categories:
