siRNA Details
Gene Symbol: Apc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106769.2 | NP_001100239.2 | 14 | 4576 | ||
XM_006240878.2 | XP_006240940.1 | 15 | 4719 | |||
XM_006240879.2 | XP_006240941.1 | 15 | 5305 | |||
XM_006240880.2 | XP_006240942.1 | 15 | 4653 | |||
XM_006240881.2 | XP_006240943.1 | 15 | 5843 | |||
XM_008765089.1 | XP_008763311.1 | 15 | 4829 |
