siRNA Details
Gene Symbol: RGD1359127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007657.1 | NP_001007658.1 | 2 | 1077 | ||
GenBank mRNA | BC079167.1 | - | 2 | 1077 | 7129744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
