siRNA Details
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199098.1 | NP_954529.1 | 3 | 610 | ||
XM_006241347.2 | XP_006241409.1 | 4 | 707 | |||
XM_006241348.2 | XP_006241410.1 | 4 | 560 | |||
XM_006241349.1 | XP_006241411.1 | 4 | 445 | |||
XM_008765355.1 | XP_008763577.1 | 3 | 656 | |||
GenBank mRNA | BC061871.1 | - | 3 | 610 | 5598899 |
Gene Symbol: LOC100912615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765371.1 | XP_008763593.1 | 715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
