siRNA Details
Gene Symbol: Cnot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011988.1 | NP_001011988.1 | 8 | 998 | ||
XM_006241363.1 | XP_006241425.1 | 10 | 1074 | |||
XM_006241364.2 | XP_006241426.1 | 10 | 1147 | |||
XM_006241365.2 | XP_006241427.1 | 11 | 1370 | |||
XM_006241366.2 | XP_006241428.1 | 11 | 1375 | |||
XM_006241367.2 | XP_006241429.1 | 11 | 1205 | |||
XM_006241369.1 | XP_006241431.1 | 9 | 951 | |||
XM_006241370.2 | XP_006241432.1 | 9 | 1080 | |||
GenBank mRNA | BC087653.1 | - | 8 | 998 | 7313082 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001037846, NM_001037847, NM_028082, AK011231, AK078121, and others.
Gene Ontology Categories:
