siRNA Details
Gene Symbol: Tbc1d31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134842.1 | NP_001128314.1 | 1, 2 | 162 | ||
XM_006241653.1 | XP_006241715.1 | 1, 2 | 92 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tbc1d31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134842.1 | NP_001128314.1 | 1, 2 | 162 | ||
XM_006241653.1 | XP_006241715.1 | 1, 2 | 92 |