siRNA Details
Gene Symbol: Ndufb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127294.2 | NP_001120766.1 | 3 | 358 | ||
GenBank mRNA | BC166927.1 | - | 3 | 358 | 6919700 | |
BC168767.1 | - | 3 | 358 | 6891022 | ||
FQ210477.1 | - | 3 | 365 | |||
FQ217145.1 | - | 3 | 364 | |||
FQ217579.1 | - | 3 | 377 | |||
FQ217728.1 | - | 3 | 358 | |||
FQ220460.1 | - | 3 | 377 | |||
FQ223542.1 | - | 3 | 365 | |||
FQ223611.1 | - | 3 | 377 | |||
FQ224000.1 | - | 3 | 423 | |||
FQ224351.1 | - | 3 | 377 | |||
FQ224618.1 | - | 3 | 394 | |||
FQ224680.1 | - | 3 | 365 | |||
FQ228493.1 | - | 3 | 365 | |||
FQ228673.1 | - | 3 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
