siRNA Details
Gene Symbol: Atp6v1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011992.1 | NP_001011992.1 | 12 | 1055 | ||
XM_006241602.2 | XP_006241664.1 | 11 | 1060 | |||
GenBank mRNA | BC089961.1 | - | 12 | 1055 | 7314200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
