siRNA Details
Gene Symbol: Rhpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241900.2 | XP_006241962.1 | 4, 5 | 555 | ||
XM_006241901.2 | XP_006241963.1 | 4, 5 | 556 | |||
XM_006241902.1 | XP_006241964.1 | 2, 3 | 169 | |||
XM_008765628.1 | XP_008763850.1 | 4, 5 | 1944 | |||
XM_216954.8 | XP_216954.4 | 4, 5 | 554 |

More Information
Other Species Targeted

Gene Ontology Categories:
