siRNA Details
Gene Symbol: Ift27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130495.1 | NP_001123967.1 | 5, 6 | 663 | ||
XM_006241931.2 | XP_006241993.1 | 5, 6 | 675 | |||
GenBank mRNA | BC168150.1 | - | 673 | 7457249 |
