siRNA Details
Gene Symbol: Gga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011994.1 | NP_001011994.1 | 8 | 915 | ||
XM_006241973.2 | XP_006242035.1 | 9 | 2277 | |||
XM_008765679.1 | XP_008763901.1 | 10 | 2033 | |||
XM_008765680.1 | XP_008763902.1 | 7 | 1531 | |||
XM_008765681.1 | XP_008763903.1 | 9 | 4739 | |||
GenBank mRNA | BC090031.1 | - | 8 | 915 | 7366311 |

More Information
Other Species Targeted

Gene Ontology Categories:
