siRNA Details
Gene Symbol: Rbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034135.1 | NP_001029307.1 | 147 | |||
GenBank mRNA | BC100258.1 | - | 147 | 7444286 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019712, BC027396, CJ082632, AK004114, BC051473, and others.
Gene Ontology Categories:
