siRNA Details
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033694.1 | NP_001028866.1 | 8 | 1559 | ||
XM_006242079.2 | XP_006242141.1 | 8 | 1914 | |||
GenBank mRNA | BC101902.1 | - | 8 | 1559 | 7934137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
