siRNA Details
Gene Symbol: Fam118a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173336.1 | NP_001166807.1 | 6, 7 | 1047 | ||
XM_006242157.2 | XP_006242219.1 | 7, 8 | 1659 | |||
XM_006242158.2 | XP_006242220.1 | 6, 7 | 1463 | |||
XM_006242159.2 | XP_006242221.1 | 6, 7 | 1386 | |||
XM_008765686.1 | XP_008763908.1 | 7, 8 | 2806 | |||
GenBank mRNA | BC169018.1 | - | 6, 7 | 1047 | 9028156 |
