siRNA Details
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130499.1 | NP_001123971.1 | 8 | 857 | ||
GenBank mRNA | FQ210197.1 | - | 8 | 826 | ||
FQ218771.1 | - | 8 | 867 |

More Information
Other Species Targeted

